EMAIL THIS PAGE TO A FRIEND
American journal of reproductive immunology (New York, N.Y. : 1989)
Latent TGF-β1 is compartmentalized between blood and seminal plasma of HIV-positive men and its activation in semen is negatively correlated with viral load and immune activation.
PMID 25052241
Abstract
Semen is the primary medium for sexual transmission of HIV-1 and contains high concentrations of TGF-β1, but its role in regulating HIV-mediated immune activation is unclear. TGF-β1 and sCD14 were compared in blood plasma (BP) and seminal plasma (SP) from HIV-uninfected and infected, antiretroviral therapy (ART)-naive and ART-treated men and in THP-1 cells following exposure to HIV-1. The relationship between TGF-β1 and sCD14 was determined by Spearman correlation. Active and latent forms of TGF-β1 were compartmentalized between BP and SP. Highest active TGF-β1 levels were present in SP of ART-naïve chronic-infected men and decreased following ART treatment. Latent TGF-β1 was upregulated in BP following HIV infection, and highest levels were observed in BP of acute-infected men. Similar expression trends were observed between latent TGF-β1 and sCD14 in BP. A significant negative correlation was observed between active TGF-β1, sCD14, and semen viral load in ART-naive men. TGF-β1 is compartmentalized between blood and semen, possibly co-expressed with sCD14 by activated monocytes/macrophages in BP as a result of HIV infection. Conversion of latent TGF-β1 into its active form could contribute to regulation of viral load and immune activation in the male genital tract, but depends on the stage of infection.