EMAIL THIS PAGE TO A FRIEND
Biodegradation
Degradation of phenanthrene by the rhizobacterium Ensifer meliloti.
PMID 25052918
Abstract
The biodegradation of the polycyclic aromatic hydrocarbon phenantherene by the rhizobacterial strain Ensifer meliloti P221, isolated from the root zone of plant grown in PAH-contaminated soil was studied. Bacterial growth and phenanthrene degradation under the influence of root-exuded organic acids were also investigated. Analysis of the metabolites produced by the strain by using thin-layer chromatography, gas chromatography, high-pressure liquid chromatography, and mass-spectrometry revealed that phenanthrene is bioconverted via two parallel pathways. The first, major pathway is through terminal aromatic ring cleavage (presumably at the C3-C4 bond) producing benzocoumarin and 1-hydroxy-2-naphthoic acid, whose further degradation with the formation of salicylic acid is difficult or is very slow. The second pathway is through the oxidation of the central aromatic ring at the C9-C10 bond, producing 9,10-dihydro-9,10-dihydroxyphenanthrene, 9,10-phenanthrenequinone, and 2,2'-diphenic acid. This is the first time that the dioxygenation of phenanthrene at the C9 and C10 atoms, proven by identification of characteristic metabolites, has been reported for a bacterium of the Ensifer genus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
116017
2-Carboxybenzaldehyde, 97%
C8H6O3