EMAIL THIS PAGE TO A FRIEND
Food chemistry
Ultra-high performance liquid chromatographic and mass spectrometric analysis of active vitamin B12 in cells of Propionibacterium and fermented cereal matrices.
PMID 25053103
Abstract
A sensitive and selective method is needed to analyse in situ produced vitamin B12 in plant-based materials, potential new dietary sources of vitamin B12. A UHPLC/UV method was developed and validated for the determination of human active vitamin B12 in cell extracts of Propionibacterium freudenreichii subsp. shermanii and after immunoaffinity purification in extracts of cereal matrices fermented by P. freudenreichii. An Acquity HSS T3 C18 column resulted in a baseline separation, a calibration curve of excellent linearity and a low limit of detection (0.075 ng/5 μL injection). As confirmed by UHPLC-MS, the active vitamin B12 could be separated from pseudovitamin B12. The recovery of vitamin B12 from purified spiked cereal matrices was good (>90%; RSD<5%). A nutritionally relevant amount of active vitamin B12 was produced by P. freudenreichii in cereal malt matrices (up to 1.9 μg/100 g) in 24h at 28 °C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL, analytical standard
C19H30O2
71251
Acetic acid, analytical standard
C2H4O2
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
409332
Cobalt(II) chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Cl2Co
449776
Cobalt(II) chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Co
C3607
Cyanocobalamin, meets USP testing specifications
C63H88CoN14O14P
43107
Cyanocobalamin, tested according to Ph.Eur.
C63H88CoN14O14P
C3000000
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
C63H88CoN14O14P
PHR1234
Cyanocobalamin, Pharmaceutical Secondary Standard; Certified Reference Material
C63H88CoN14O14P
BP466
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
C63H88CoN14O14P
47869
Cyanocobalamin (B12), analytical standard
C63H88CoN14O14P
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2
71430
Sodium cyanide, purum p.a., ≥96.0% (AT)
CNNa
205222
Sodium cyanide, ACS reagent, ≥95.0%
CNNa
380970
Sodium cyanide, reagent grade, 97%
CNNa
43617
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
HNaO
72064
Sodium hydroxide solution, 50-52% in water, eluent for IC
HNaO
72071
Sodium hydroxide solution, for the determination of nitrogen, ≥32.0% in H2O (T)
HNaO
74564
Trifluoroacetic acid, analytical standard
C2HF3O2
V2876
Vitamin B12, ≥98%
C63H88CoN14O14P
V6629
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
C63H88CoN14O14P