EMAIL THIS PAGE TO A FRIEND
Immunobiology
Cholesterol crystal-induced endothelial cell activation is complement-dependent and mediated by TNF.
PMID 25053140
Abstract
Cholesterol crystals are known to be a hallmark of atherosclerosis with recent studies demonstrating deposition of these crystals in early fatty streak formation as well as penetrating the intima following plaque rupture. Inflammation has also become a central focus in atheroma development and endothelial cell activation is recognized as necessary for the recruitment of inflammatory cells to the plaque. However, the extent to which cholesterol crystals can induce inflammation and activate endothelial cells is not known. To investigate this, we developed a novel model activating human umbilical vein endothelial cells using lepirudin anticoagulated human whole blood. We found that cholesterol crystals caused a marked and dose-dependent increase in the adhesion molecules E-selectin and ICAM-1 on the surface of the endothelial cells after incubation with whole blood. There was no activation of the cells when the crystals were incubated in medium alone, or in human serum, despite substantial crystal-induced complement activation in serum. Complement inhibitors at the C3 and C5 levels reduced the whole blood induced endothelial cell activation by up to 89% (p<0.05) and abolished TNF release (p<0.01). Finally, the TNF inhibitor infliximab reduced endothelial activation to background levels (p<0.05). In conclusion, these data demonstrate that endothelial activation by cholesterol crystals is mediated by complement-dependent TNF release, and suggests that complement-inhibition might have a role in alleviating atherosclerosis-induced inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405580
Anti-MS4A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014341
Anti-MS4A1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA014391
Anti-MS4A1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502982
Anti-TNFA antibody produced in rabbit, affinity isolated antibody
T0813 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T1816 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T0938 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T8300
Anti-Tumor Necrosis Factor-α antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4700114 Monoclonal Anti-CD20 antibody produced in mouse, clone 2H7, purified immunoglobulin, buffered aqueous solution
SAB4700119 Monoclonal Anti-CD20 antibody produced in mouse, clone MEM-269, purified immunoglobulin, buffered aqueous solution
SAB4700121
Monoclonal Anti-CD20 antibody produced in mouse, clone MEM-97, purified immunoglobulin, buffered aqueous solution
WH0000931M1
Monoclonal Anti-MS4A1 antibody produced in mouse, clone 5C11, purified immunoglobulin, buffered aqueous solution
SAB1403646
Monoclonal Anti-MS4A1 antibody produced in mouse, clone 4F11, purified immunoglobulin, buffered aqueous solution
WH0007124M2
Monoclonal Anti-TNF antibody produced in mouse, clone 1C3-A1-F4, purified immunoglobulin, buffered aqueous solution
SAB1404480
Monoclonal Anti-TNF antibody produced in mouse, clone M1-C4, purified immunoglobulin, buffered aqueous solution
T3198 Monoclonal Anti-Tumor Necrosis Factor-α antibody produced in mouse, clone 45418.111, purified immunoglobulin, lyophilized powder
T6817 Monoclonal Anti-Tumor Necrosis Factor-α antibody produced in mouse, clone 28401, purified immunoglobulin, lyophilized powder
SRP3177
TNF-α human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
T6674
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
H8916
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture