EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Concerns over data in key dabigatran trial.
PMID 25055831
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1349
Beta Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2