EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Complement-mediated glomerular injury is reduced by inhibition of protein-tyrosine phosphatase 1B.
PMID 25056348
Abstract
The unfolded protein response and endoplasmic reticulum-associated degradation (ERAD) contribute to injury in renal glomerular diseases, including those mediated by complement C5b-9. In the present study, we address the role of protein-tyrosine phosphatase 1B (PTP1B) in complement-mediated glomerular injury and ERAD. In glomerular epithelial cells (GECs)/podocytes and PTP1B-deficient mouse embryonic fibroblasts exposed to complement, inhibition/deletion of PTP1B reduced ERAD, as monitored by the ERAD reporter CD3δ. Overexpression of PTP1B produced an effect similar to PTP1B deficiency on ERAD in complement-treated GECs. Complement-mediated cytotoxicity was reduced after PTP1B overexpression and tended to be reduced after PTP1B inhibition. PTP1B enhanced the induction of certain ERAD components via the inositol-requiring-1α branch of the unfolded protein response. PTP1B knockout mice with anti-glomerular basement membrane glomerulonephritis had decreased proteinuria and showed less podocyte loss and endoplasmic reticulum dysfunction compared with wild-type littermates. These results imply that endogenous levels of PTP1B are tightly regulated and that both overexpression and inhibition can affect ERAD. The cytoprotective effects of PTP1B deletion in cultured cells and in anti-glomerular basement membrane nephritis suggest that PTP1B may potentially be a therapeutic target in complement-mediated diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151645
Thiazole, 99%
C3H3NS
W361518
Thiazole, ≥99%
C3H3NS