EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Genetic removal of matrix metalloproteinase 9 rescues the symptoms of fragile X syndrome in a mouse model.
PMID 25057190
Abstract
Fmr1 knock-out (ko) mice display key features of fragile X syndrome (FXS), including delayed dendritic spine maturation and FXS-associated behaviors, such as poor socialization, obsessive-compulsive behavior, and hyperactivity. Here we provide conclusive evidence that matrix metalloproteinase-9 (MMP-9) is necessary to the development of FXS-associated defects in Fmr1 ko mice. Genetic disruption of Mmp-9 rescued key aspects of Fmr1 deficiency, including dendritic spine abnormalities, abnormal mGluR5-dependent LTD, as well as aberrant behaviors in open field and social novelty tests. Remarkably, MMP-9 deficiency also corrected non-neural features of Fmr1 deficiency-specifically macroorchidism-indicating that MMP-9 dysregulation contributes to FXS-associated abnormalities outside the CNS. Further, MMP-9 deficiency suppressed elevations of Akt, mammalian target of rapamycin, and eukaryotic translation initiation factor 4E phosphorylation seen in Fmr1 ko mice, which are also associated with other autistic spectrum disorders. These findings establish that MMP-9 is critical to the mechanisms responsible for neural and non-neural aspects of the FXS phenotype.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA010122
Anti-DLG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401115
Anti-DLG4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
06-543
Anti-FAK Antibody, clone, Upstate®, from rabbit
SAB4502498
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502495
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502499
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502496
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502503
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4200232
Anti-GAD65 antibody , Mouse monoclonal, clone N-GAD65, purified from hybridoma cell culture
SAB4300597
Anti-PTK2 (Ab-576/577) antibody produced in rabbit, affinity isolated antibody
SAB4300355
Anti-PTK2 (Ab-861) antibody produced in rabbit, affinity isolated antibody
SAB4300418
Anti-PTK2 (Ab-925) antibody produced in rabbit, affinity isolated antibody
SAB4300513
Anti-SYN1 (Ab-9) antibody produced in rabbit, affinity isolated antibody
HPA000397
Anti-SYN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AB1543P
Anti-Synapsin I Antibody, clone, Chemicon®, from rabbit
S193 Anti-Synapsin I antibody produced in rabbit, affinity isolated antibody, lyophilized powder
G5384 Gelatin-Agarose, saline suspension
RAB0466
Human TIMP-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0467
Human TIMP-1 ELISA Kit, for cell and tissue lysates
G1166 Monoclonal Anti-Glutamic Acid Decarboxylase 65 antibody produced in mouse, clone GAD-6, purified immunoglobulin, buffered aqueous solution
RAB0468
Mouse TIMP-1 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0469
Mouse TIMP-1 ELISA Kit, for cell and tissue lysates
P1406 Protein A–Agarose, lyophilized powder
RAB0470
Rat TIMP-1 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0471
Rat TIMP-1 ELISA Kit, for cell and tissue lysates
SAB4300665
Anti-FAK (Ab-397) antibody produced in rabbit, affinity isolated antibody
SAB2105773
Anti-DLG4 antibody produced in rabbit, affinity isolated antibody
SAB1405839
Anti-GAD2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution