EMAIL THIS PAGE TO A FRIEND
The Journal of Clinical Endocrinology and Metabolism
5-Hydroxymethylcytosine promotes proliferation of human uterine leiomyoma: a biological link to a new epigenetic modification in benign tumors.
PMID 25057885