EMAIL THIS PAGE TO A FRIEND
Pharmacology, Biochemistry, and Behavior
Combined inhibition of FAAH and COX produces enhanced anti-allodynic effects in mouse neuropathic and inflammatory pain models.
PMID 25058512