EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
In vitro antitrypanosomal and antiplasmodial activities of crude extracts and essential oils of Ocimum gratissimum Linn from Benin and influence of vegetative stage.
PMID 25058875
Abstract
Different parts of Ocimum gratissimum Linn are largely used in folk medicine for the treatment of many diseases, some of which related to parasitical infections as fevers and headaches. In order to validate their use and to clarify the plant part which possesses the best antiparasitic properties, we decided to evaluate the in vitro antiplasmodial and antitrypanosomal activities of essential oils and crude extracts from leaves, stems and seeds of Ocimum gratissimum as well as their cytotoxicity. The essential oils and ethanol crude extracts of leaves and stems of Ocimum gratissimum from Benin, were obtained in pre and full flowering stages. Seeds obtained only in full flowering stage, were also extracted. The oils were isolated by hydrodistillation and analyzed by GC/MS and GC/FID. Extracts and essential oils were tested in vitro against Trypanosoma brucei brucei and Plasmodium falciparum. Cytotoxicity was evaluated in vitro against Chinese Hamster Ovary (CHO) cells and the human non cancer fibroblast cell line (WI38) through MTT assay to evaluate the selectivity and toxicity was assessed against Artemia salina Leach. The essential oils and non-volatile crude extracts of Ocimum gratissimum were more active on Trypanosoma brucei brucei than on Plasmodium falciparum (3D7). This activity varies according to the vegetative stage (pre and full flowering) and the plant part (seeds, stems and leaves) extracted. The best growth inhibition of Trypanosoma brucei brucei was observed with ethanol crude extracts of leaves (IC50=1.66 ± 0.48 μg/mL) and seeds (IC50=1.29 ± 0.42 μg/mL) in full flowering stage with good selectivity (SI>10). The chemical composition of the essential oil from aerial parts (47 compounds), characterized by the presence as main constituents of p-cymene, thymol, γ-terpinene, β-myrcene and α-thujene, depends on the vegetative stage. The oil contained some minor compounds such as myrcene (IC50=2.24 ± 0.27μg/mL), citronellal (IC50=2.76 ± 1.55μg/mL), limonene (IC50=4.24 ± 2.27μg/mL), with good antitrypanosomal activities. These oils and crude extracts were not toxic against Artemia salina Leach and had a low cytotoxicity except leaves and seeds ethanol extracts obtained in full flowering which showed toxicity against CHO and WI38 cells. Our study shows that ethanol crude extracts of leaves and seeds of Ocimum gratissimum in full flowering stage can be a good source of antitrypanosomal agents. This is the first report about the relation between the plant part extracted, the vegetative stage of the plant, the antitrypanosomal and antiplasmodial activities and the cytotoxicity of essential oils and non-volatile extracts of Ocimum gratissimum from Benin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20506
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
C10H16O
21310
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
C10H16O
W526606
(±)-Camphor, ≥95.5%
C10H16O
21300
(+)-Camphor, purum, natural, ≥97.0% (sum of enantiomers, GC)
C10H16O
45855
(−)-Bornyl acetate, analytical standard
C12H20O2
B55203
(−)-Bornyl acetate, 95%
C12H20O2
W408001
(−)-Bornyl acetate, 95%
C12H20O2
21293
(−)-Camphor, analytical standard
C10H16O
21295
(−)-Camphor, purum, ≥95.0% (sum of enantiomers, GC)
C10H16O
361593
Artemisinin, 98%
C15H22O5
456055
Camphene, 95%
C10H16
01410590
Camphene, primary reference standard
C10H16
442505
Camphene, analytical standard
C10H16
148075
Camphor, 96%
C10H16O
1087508
Camphor, United States Pharmacopeia (USP) Reference Standard
C10H16O
295299
Dimethyl ether, ≥99%
C2H6O
W250716
Geraniol, natural, ≥97%, FG
C10H18O
W250708
Geraniol, ≥97%, FCC, FG
C10H18O
48798
Geraniol, analytical standard
C10H18O
163333
Geraniol, 98%
C10H18O
42583
Lavandulol, analytical standard
C10H18O
W276200
Myrcene, stabilized, FCC, FG
C10H16
64643
Myrcene, analytical standard
C10H16
M100005
Myrcene, technical grade
C10H16
04600585
Myrcene, primary reference standard
C10H16
C121452
p-Cymene, 99%
C10H14
W235601
p-Cymene, ≥97%, FG
C10H14
30039
p-Cymene, analytical standard
C10H14
49679
p-Cymene, certified reference material, TraceCERT®
C10H14
W304603
Terpinolene, ≥90%
C10H16