EMAIL THIS PAGE TO A FRIEND
Talanta
Speciation of As(III)/As(V) in water samples by a magnetic solid phase extraction based on Fe?O?/Mg-Al layered double hydroxide nano-hybrid followed by chemiluminescence detection.
PMID 25059142