EMAIL THIS PAGE TO A FRIEND
Clinical chemistry and laboratory medicine
Quantification of adult T-cell leukemia/lymphoma cells using simple four-color flow cytometry.
PMID 25060346
Abstract
The absolute number of adult T-cell leukemia/lymphoma (ATL) cells in peripheral blood is an essential indicator to evaluate disease status. However, microscopically counting ATL cells based on morphology requires experience and tends to be inaccurate due to the rarity of ATL. Based on our research showing that acute-type ATL cells are specifically enriched in the CD4+/CD7- (CD7N) fraction, a new analytical method to accurately quantify ATL cells was established using an internal bead standard and simple four-color flow cytometry. This method was verified by comparison with microscopic examination of 49 peripheral blood samples and used to follow up patients. A strong correlation was observed between the number of CD7N cells measured by flow cytometry and the number of abnormal lymphocytes measured microscopically by experienced technicians [Pearson's R, 0.963; Spearman's rho, 0.921; intercorrelation coefficient, 0.962]. The linear regression coefficient was close to 1 (β=1.013). Our method could detect 1 cell/μL, and the limit of quantitation was between 2.9 and 9.8 cells/μL. The frequency of CD7N cells among CD4+ cells changed during chemotherapy, which reflected differences between chemosensitive and chemoresistant cases. Kaplan-Meier analysis with a log-rank test showed that patients with decreased CD7N proportion after chemotherapy had significantly longer disease-specific survival (p=0.003). Our newly established method quantified tumor cells in patients with acute-type ATL. Furthermore, this method was useful for assessing the efficacy of chemotherapy, and the change of the CD7N proportion could be more important to predict prognosis.