EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Effective 2D-RPLC/RPLC enrichment and separation of micro-components from Hedyotis diffusa Willd. and characterization by using ultra-performance liquid chromatography/quadrupole time-of-flight mass spectrometry.
PMID 25061712
Abstract
An effective method aiming at enrichment and analysis of micro-components in traditional Chinese medicine (TCM) was developed. One fraction (fraction E) from the extract of Hedyotis diffusa Willd. was selected as test sample, which was isolated by using the XAD-4 macroporous resin. To study the micro-components, a two-dimensional reverse-phase liquid chromatography (2D-RPLC/RPLC) method was developed, comprising Click OEG and C18 stationary phases as the first and second dimensions, respectively. Of the eight sub-fractions isolated from the first dimension, three sub-fractions (fractions II-IV) containing micro-components were further separated with the second dimension. The 2D-RPLC/RPLC system was proved to possess high orthogonality. Furthermore, the micro-components were characterized by using ultra-performance liquid chromatography-diode array detector/quadrupole time-of-flight mass spectrometry (UPLC-DAD/Q-TOF MS) with electrospray ionization (ESI) source. With the optimized separation and characterization method, a large number (>400) of micro-components were enriched and detected from the extracts of H. diffusa Willd., the majority of which has not been isolated from the herb before. Among these isolated micro-components, 38 compounds involving 24 phenylpropanoids, 7 flavonoids and 7 iridoid glucosides (IGs), were identified or tentatively identified from the H. diffusa extracts on the basis of spectral data of the authentic standards and the fragmentation characteristics information available in literatures. The proposed method made it possible to effectively screen and analyze the micro-components in TCMs or other complex natural medicines.