EMAIL THIS PAGE TO A FRIEND
Domestic Animal Endocrinology
Vascular endothelial growth factor A improves quality of matured porcine oocytes and developing parthenotes.
PMID 25061966
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300356
Anti-KDR (Ab-1175) antibody produced in rabbit, affinity isolated antibody
SAB4300357
Anti-KDR (Ab-1214) antibody produced in rabbit, affinity isolated antibody
SAB4300358
Anti-KDR (Ab-951) antibody produced in rabbit, affinity isolated antibody
SAB2900238
Anti-KDR (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900239
Anti-KDR (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101239
Anti-KDR antibody produced in rabbit, affinity isolated antibody
V1014 Anti-Vascular Endothelial Growth Factor Receptor 2 antibody produced in goat, affinity isolated antibody, lyophilized powder
GW21181 Anti-VEGFR-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300657
Anti-VEGFR2 (Ab-1059) antibody produced in rabbit, affinity isolated antibody
SAB4501645
Anti-VEGFR2 antibody produced in rabbit, affinity isolated antibody
WH0003791M1
Monoclonal Anti-KDR antibody produced in mouse, clone 2A2, purified immunoglobulin, buffered aqueous solution
SAB1404004
Monoclonal Anti-KDR antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
V9134
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse, clone KDR-1, ascites fluid
V7259 Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
SRP3182
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested