EMAIL THIS PAGE TO A FRIEND
BMC Complementary and Alternative Medicine
Effects of Korean red ginseng (Panax Ginseng Meyer) on bisphenol A exposure and gynecologic complaints: single blind, randomized clinical trial of efficacy and safety.
PMID 25063041