EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Meta-analyses of Dehalococcoides mccartyi strain 195 transcriptomic profiles identify a respiration rate-related gene expression transition point and interoperon recruitment of a key oxidoreductase subunit.
PMID 25063656