EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Biofilm formation on biotic and abiotic surfaces in the presence of antimicrobials by Escherichia coli Isolates from cases of bovine mastitis.
PMID 25063668