EMAIL THIS PAGE TO A FRIEND
Free Radical Biology & Medicine
Glutathione sulfinamide serves as a selective, endogenous biomarker for nitroxyl after exposure to therapeutic levels of donors.
PMID 25064322