EMAIL THIS PAGE TO A FRIEND
Gene
Rgn gene is required for gut cell homeostasis after ingestion of sodium dodecyl sulfate in Drosophila.
PMID 25064488
Abstract
Resistance and resilience constitute the two complementary aspects of epithelial host defenses in Drosophila. Epithelial cell homeostasis is necessary for the recovery of damages caused by stress or infections. However, the genes responsible for gut epithelial homeostasis remain poorly understood. Here, we show that rgn(G4035) mutant flies have higher mortality than wild-type flies after ingestion of sodium dodecyl sulfate (SDS). Excessive melanization and increased necrotic cells in the gut contribute to the reduced survival of rgn(G4035) mutant flies following SDS ingestion. rgn mutant flies have a defect in the replenishment of intestinal stem cells (ISCs) following gut damage. The antimicrobial peptide (AMP) expression is affected in rgn(G4035) mutant fly guts. Together, our study provides evidence that rgn gene is essential for gut cell homeostasis following damage in Drosophila.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6830 Cecropin A, ≥97% (HPLC), powder
C184H313N53O46