EMAIL THIS PAGE TO A FRIEND
FEBS letters
Dynein-dependent transport of spindle assembly checkpoint proteins off kinetochores toward spindle poles.
PMID 25064841
Abstract
A predominant mechanism of spindle assembly checkpoint (SAC) silencing is dynein-mediated transport of certain kinetochore proteins along microtubules. There are still conflicting data as to which SAC proteins are dynein cargoes. Using two ATP reduction assays, we found that the core SAC proteins Mad1, Mad2, Bub1, BubR1, and Bub3 redistributed from attached kinetochores to spindle poles, in a dynein-dependent manner. This redistribution still occurred in metaphase-arrested cells, at a time when the SAC should be satisfied and silenced. Unexpectedly, we found that a pool of Hec1 and Mis12 also relocalizes to spindle poles, suggesting KMN components as additional dynein cargoes. The potential significance of these results for SAC silencing is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1411705
Anti-CDC20 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2103238
Anti-CDC20, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4503731
Anti-p55CDC antibody produced in rabbit, affinity isolated antibody
SAB2700526
Anti-SS18L1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700532
Anti-SS18L1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B0561 Monoclonal Anti-BUB1 antibody produced in mouse, 2 mg/mL, clone 14H5, purified immunoglobulin, buffered aqueous solution
WH0000699M1
Monoclonal Anti-BUB1 antibody produced in mouse, clone 4C3-4C9, purified immunoglobulin, buffered aqueous solution
SAB1403620
Monoclonal Anti-BUB1, (N-terminal) antibody produced in mouse, clone 2F9, purified immunoglobulin, buffered aqueous solution
D5167 Monoclonal Anti-Dynein (Intermediate Chain) antibody produced in mouse, clone 70.1, ascites fluid
74540
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
C18H22O4
N5023
Nordihydroguaiaretic acid, ≥90% (HPLC), from Larrea divaricata (creosote bush)
C18H22O4
PLA0173
Rabbit anti-CDC20 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB2103841
Anti-SS18L1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1304653
ANTI-SS18L1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution