EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Lichenysin, a cyclooctapeptide occurring in Chinese liquor jiannanchun reduced the headspace concentration of phenolic off-flavors via hydrogen-bond interactions.
PMID 25065507
Abstract
Nonvolatile compounds play important roles in the quality of alcoholic beverages. In our previous work, a type of cyclooctapeptide lichenysin was newly identified in Chinese strong-aroma type liquor. In this work, it was found that lichenysin could selectively affect aroma volatility in strong-aroma type (Jiannanchun) liquor. Interaction of lichenysin and volatile phenolic compounds (off-odors in strong-aroma type liquor) was characterized using headspace solid-phase microextraction coupled with gas chromatography-mass spectrometry (HS-SPME-GC-MS). HS-SPME results indicated that lichenysin very efficiently suppressed the volatility of phenolic compounds by 36-48% (P < 0.05). Thermodynamic analysis showed that the binding process was mainly mediated by hydrogen bonding. Furthermore, the mixture of lichenysin and 4-ethylguaiacol revealed intermolecular cross peaks between the aH (Val) of lichenysin and the 1H of 4-ethylguaiacol, by using nuclear Overhauser effect spectroscopy. This study will help to further understand the interaction mechanisms between flavor and nonvolatile matrix components in Chinese liquors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74858
(±)-2-Octanol, ReagentPlus®, ≥99.5% (GC)
C8H18O
42277
(±)-2-Octanol, analytical standard
C8H18O
74860
(±)-2-Octanol, ≥96.0% (GC)
C8H18O
O4504
2-Octanol, 97%
C8H18O
W280100
2-Octanol, ≥97%
C8H18O
W280110
2-Octanol, natural, ≥97%, FG
C8H18O
156914
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
453323
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
424005
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
417939
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
417920
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 1 % (v/v) TMS
C2D6OS
570672
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
C2D6OS
569585
Dimethyl sulfoxide-d6, 99.9 atom % D, anhydrous
C2D6OS
151874
Dimethyl sulfoxide-d6, 99.9 atom % D
C2D6OS
522058
Dimethyl sulfoxide-d6, 99.9 atom % D
C2D6OS
612324
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
C2D6OS
716731
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
C2D6OS
296147
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
574341
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
185965
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
C2D6OS
175943
Dimethyl sulfoxide-d6, 99.5 atom % D
C2D6OS
724335
Dimethyl sulfoxide-d6, contains 0.07 wt. % TSP-d4, 99.9 atom % D, 99% (CP)
C2D6OS
W257001
Hexyl formate, ≥97%, FG
C7H14O2
87920
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
C4H12Si
87921
Tetramethylsilane, ≥99.0% (GC)
C4H12Si
744301
Tetramethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C4H12Si
523771
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
C4H12Si
T24007
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
C4H12Si