EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Studies on the reaction of trans-2-heptenal with peanut proteins.
PMID 25065678
Abstract
Hexanal, 2-heptenal, and nonanal were identified as relevant reaction products formed in the course of the lipid peroxidation of heated peanut oil. For the identification of potential amino acid side chain adducts, kinetic studies between N(α)-benzoylglycyl-l-lysine as a model for protein-bound lysine and trans-2-heptenal were performed, showing a strong decrease of the lysine-derivative whereupon the loss of trans-2-heptenal was moderate. Following acid hydrolysis of the incubation mixture of N(α)-acetyl-l-lysine and trans-2-heptenal, two UV-active major lipation products were observed, isolated and identified as isomeric pyridinium-derivatives, namely (Z)- and (E)-1-(5-amino-5-carboxypentyl)-4-butyl-3-(pent-1-en-1-yl)pyridin-1-ium (cis- and trans-BPP-lysine). After heating of a native peanut protein extract with trans-2-heptenal, both derivatives were quantitated by LC-ESI-MS/MS after acid hydrolysis and the modification of lysine was measured by amino acid analysis. At low, "food-relevant", concentrations of trans-2-heptenal, up to 80% of the lysine modification could be explained by the formation of cis- and trans-BPP-lysine, showing that these two lipation derivatives represent good markers for a protein modification by the lipid peroxidation product trans-2-heptenal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
97865
1,3-Diethylbenzene, ≥98.0% (GC)
C10H14
105899
Cyclohexanol, ReagentPlus®, 99%
C6H12O
44113
Cyclohexanol, analytical standard
C6H12O
61696
Heptaldehyde, analytical standard
C7H14O
H2120
Heptaldehyde, 95%
C7H14O
W254002
Heptanal, ≥95%, FCC, FG
C7H14O
115606
Hexanal, 98%
C6H12O
W255718
Hexanal, ≥97%, FCC, FG
C6H12O
W255726
Hexanal, natural, ≥95%, FG
C6H12O
18109
Hexanal, analytical standard
C6H12O
57180
Indo 1, for fluorescence, ≥80.0% (HPLC)
C32H26K5N3O12
194166
Methanol-d4, "100%", ≥99.96 atom % D, ≥99% (CP)
CD4O
721948 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 5 mm × 8 in.
CD4O
721921 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 3 mm × 8 in.
CD4O
444758
Methanol-d4, "100%", 99.96 atom % D
CD4O
535435
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
CD4O
151947
Methanol-d4, ≥99.8 atom % D
CD4O
441384
Methanol-d4, ≥99.8 atom % D
CD4O
343803
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
530530
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
611646
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
CD4O
439029
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
CD4O
417653
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
CD4O
422878
Methanol-d4, 99 atom % D
CD4O
569534
Methanol-d4, anhydrous, ≥99.8 atom % D
CD4O
570729
Methanol-d4, ≥99.8 atom % D, anhydrous
CD4O
W278203
Nonanal, ≥95%, FCC
C9H18O
N30803
Nonanal, 95%
C9H18O
W278220
Nonanal, natural, ≥98%, FG
C9H18O
442719
Nonanal, analytical standard
C9H18O