EMAIL THIS PAGE TO A FRIEND
Chemosphere
Improved arsenic(III) adsorption by Al2O3 nanoparticles and H2O2: evidence of oxidation to arsenic(V) from X-ray absorption spectroscopy.
PMID 25065803
Abstract
We have investigated the oxidation of inorganic As(III) with H2O2 catalysed by Al2O3, using X-ray absorption near-edge structure and extended X-ray absorption fine structure spectroscopy. The effects of different reaction conditions (pH, time and initial H2O2 concentration) were also studied as were the kinetics of the oxidation reaction. We demonstrated that As(III) was oxidized to As(V) in the presence of H2O2 and Al2O3. Furthermore, all arsenic species found on the Al2O3 surface were in the As(V) state. The presence of both Al2O3 and H2O2 was necessary for oxidation of As(III) to take place within the period of time studied. The oxidation kinetics indicate a mechanism where reversible As(III) binding to the alumina surface is followed by irreversible oxidation by H2O2 leading to strongly bound As(V). Results from this study indicate that there is a surface-catalysed oxidation of As(III) on Al2O3 by H2O2, a reaction that can take place in nature and can be of help in the development of novel treatment systems for As(III) removal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199443
Aluminum oxide, activated, basic, Brockmann I
Al2O3
199974
Aluminum oxide, activated, neutral, Brockmann I
Al2O3
199966
Aluminum oxide, activated, acidic, Brockmann I
Al2O3
229423
Aluminum oxide, powder, 99.99% trace metals basis
Al2O3
265497
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Al2O3
414069
Aluminum oxide, pellets, 3 mm
Al2O3
234745
Aluminum oxide, Corundum, α-phase, -100 mesh
Al2O3
267740
Aluminum oxide, pore size 58 Å, ~150 mesh
Al2O3
A1522
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Al2O3
342688
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Al2O3
342718
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Al2O3
342653
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Al2O3
799319
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Al2O3
799300
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Al2O3
216763
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
H2O2
516813
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
H2O2
323381
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
H2O2
H3410
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
H2O2
349887
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
H2O2
95299
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
H2O2
31642
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
H2O2