EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Rapid quantitative chiral amphetamines liquid chromatography-tandem mass spectrometry: method in plasma and oral fluid with a cost-effective chiral derivatizing reagent.
PMID 25065924