EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Unconventional pore and defect generation in molybdenum disulfide: application in high-rate lithium-ion batteries and the hydrogen evolution reaction.
PMID 25066369