EMAIL THIS PAGE TO A FRIEND
Journal of Veterinary Pharmacology and Therapeutics
Pharmacokinetics and pharmacodynamics of intravenous dexmedetomidine in the horse.
PMID 25066475
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0956 Dexmedetomidine hydrochloride, ≥98% (HPLC)
C13H16N2 · HCl