EMAIL THIS PAGE TO A FRIEND
Journal of Veterinary Pharmacology and Therapeutics
Antioxidant activity of hyaluronic acid investigated by means of chemiluminescence of equine neutrophil bursts and electron paramagnetic resonance spectroscopy.
PMID 25066541