EMAIL THIS PAGE TO A FRIEND
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society
Enhanced solubility and intestinal absorption of candesartan cilexetil solid dispersions using everted rat intestinal sacs.
PMID 25067902