EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Erythropoiesis-stimulating agents and quality of life: personal journeys of a cancer survivor, oncologist, and two cancer health services researchers.
PMID 25072302