EMAIL THIS PAGE TO A FRIEND
Nutrition and Cancer
Water extract from Pleurotus pulmonarius with antioxidant activity exerts in vivo chemoprophylaxis and chemosensitization for liver cancer.
PMID 25072857