EMAIL THIS PAGE TO A FRIEND
Clinical Nuclear Medicine
A frequency and semiquantitative analysis of pathological 68Ga DOTATATE PET/CT uptake by primary site-dependent neuroendocrine tumor metastasis.
PMID 25072928