EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Pharmacokinetic study of HS061, a new human insulin, in non-diabetic rat using ultra performance liquid chromatography-tandem mass spectrometry.
PMID 25073106