EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Stimulation of the methylcoenzyme M reduction by uridine-5'-diphospho-sugars in cell-free extracts of Methanobacterium thermoautotrophicum (strain delta H).
PMID 2507320
Abstract
The hydrogen-dependent reduction of methylcoenzyme M catalyzed by coenzyme-depleted cell-free extracts of Methanobacterium thermoautotrophicum was stimulated by micromolar concentrations of a UDP-disaccharide present in the organism. The compound was isolated and identified as UDP-1-O-alpha-D-2-acetamido-2-deoxyglucopyranose (UDPGlcpNAc) glycosidically linked to 2-acetamido-2-deoxymannopyranosyluronic acid. Maximal stimulation was observed when both the UDP-disaccharide and mercaptoheptanoylthreonine phosphate were present in the reaction mixtures. The UDP derivative isolated was not specific in its action: other UDP-sugars tested in micromolar concentrations stimulated the methylcoenzyme M reduction to the same extent. The activated sugars presumably substitute for ATP, which is usually required in much higher concentrations to activate the methylcoenzyme M reductase system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50187
Ammonium 2-(methylthio)ethanesulfonate, ≥97.0% (TLC)
C3H11NO3S2