EMAIL THIS PAGE TO A FRIEND
Journal of neurology, neurosurgery, and psychiatry
Effective treatment with intravenous immunoglobulins reduces autoreactive T-cell response in patients with CIDP.
PMID 25074566
Abstract
To investigate changes in autoreactive T-cell responses against PMP-22 and P2 antigen as well as a T-cell memory repertoire in patients with chronic inflammatory demyelinating polyneuropathy (CIDP) induced by repeated intravenous immunoglobulin (IVIg) treatment. In an observational trial, we prepared cryopreserved human peripheral blood monocytes from blood from 34 patients with CIDP (18 treatment naïve and 16 maintenance IVIg treatment) and from 14 healthy controls (non-immune neuropathy and healthy control). Treatment response was defined by clinical evaluation. The autoantigen-specific T-cell response was analysed by enzyme linked immunosorbent spot (ELISPOT) assay before IVIg start (baseline) and at follow-up. The T-cell memory subsets were analysed by using flow cytometric analysis. Myelin-derived P2-specific and PMP-22-specific IFN-γ producers were increased in IVIg responders compared with non-responders before treatment, which decreased by repeated IVIg infusion cycles. Treatment responders but not non-responders showed higher frequencies of CD4 T effector memory (TEM) and T central memory frequencies at baseline compared with maintenance IVIg treatment patients and controls. In addition, IVIg treatment was associated with a significant reduction in CD8 TEM at follow-up. Our data demonstrate that immunomodulatory treatment with IVIgs on a long-term basis reduces the autoreactive T-cell response against PMP-22 and P2-antigens, which may be influenced by the altered maintenance of CD8 and CD4 effector/memory T-cell subsets towards a more anti-inflammatory immune status. Elevated PMP-22 and P2-specific T-cell responses may serve as predictors for treatment responsiveness to IVIgs warranting validation in larger studies.