EMAIL THIS PAGE TO A FRIEND
Mechanisms of Ageing and Development
Short-term rapamycin treatment in mice has few effects on the transcriptome of white adipose tissue compared to dietary restriction.
PMID 25075714