EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Immunomodulatory effect of a wild blueberry anthocyanin-rich extract in human Caco-2 intestinal cells.
PMID 25075866
Abstract
Intestinal inflammation is a natural process crucial for the maintenance of gut functioning. However, abnormal or prolonged inflammatory responses may lead to the onset of chronic degenerative diseases, typically treated by means of pharmacological interventions. Dietary strategies for the prevention of inflammation are a safer alternative to pharmacotherapy. Anthocyanins and other polyphenols have been documented to display anti-inflammatory activity. In the present study, three bioactive fractions (anthocyanin, phenolic, and water-soluble fractions) were extracted from a wild blueberry powder. The Caco-2 intestinal model was used to test the immunomodulatory effect of the above fractions. Only the anthocyanin-rich fraction reduced the activation of NF-κB, induced by IL-1β in intestinal epithelial Caco-2 cells. Specifically, concentrations of 50 and 100 μg mL(-1) decreased NF-κB activation by 68.9 and 85.2%, respectively (p ≤ 0.05). These preliminary results provide further support for the role of food bioactives as potential dietary anti-inflammatory agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
38730 Citrate standard for IC, 1000 mg/L, analytical standard
C6H8O7
94676
Citric acid, certified reference material, TraceCERT®
C6H8O7
46933
Citric acid, analytical standard
C6H8O7
1134368
Citric acid, United States Pharmacopeia (USP) Reference Standard
C6H8O7
A1202000
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
C6H8O7
PHR1071
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
C6H8O7
F0127
D-(−)-Fructose, ≥99%
C6H12O6
F2543
D-(−)-Fructose, BioXtra
C6H12O6
F3510
D-(−)-Fructose, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H12O6
F9048
D-(−)-Fructose, meets USP testing specifications
C6H12O6
F2793
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
C6H12O6
47739
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
C6H12O6
47740
D-(−)-Fructose, for microbiology, ≥99.0%
C6H12O6
47748
D-(−)-Fructose, tested according to Ph.Eur.
C6H12O6
58958
Ethyl acetate, analytical standard
C4H8O2
17023
Ethyl acetate, tested according to Ph.Eur.
C4H8O2
PHR1481
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O2
1286504
Fructose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
F0550000
Fructose, European Pharmacopoeia (EP) Reference Standard
C6H12O6
PHR1002
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6