EMAIL THIS PAGE TO A FRIEND
Yi chuan = Hereditas
Response of maize serine/arginine-rich protein gene family in seedlings to drought stress.
PMID 25076035
Abstract
Alternative splicing (AS) in eukaryotic organisms is closely related to the gene regulation in plant abiotic stress responses, in which serine/arginine-rich proteins (SR proteins) act as key regulators. The genome sequence of maize inbred line B73 was analyzed, showing that the promoter regions of SR genes possess about three to eight kinds of cis-acting regulatory elements. Twenty-seven SR genes encode alkaline proteins, and 23 of which are divided into five subgroups in terms of the first RNA recognition motif (RRM) at the amino terminal. The expression of SR genes showed tissue-specific and genotype-dependent features under drought stress in the hybrid Zhengdan-958 and its parents, Zheng-58 and Chang-7-2 via bidirectional hierarchical clustering. SR genes were down-regulated in roots while they were up-regulated in shoots under drought stress. However, SR genes were down-regulated in both roots and shoots in three different rehydration stages after severe drought stress. Additionally, a widespread alternative splicing exists in all SR genes although SR genes showed differential expression tendency under drought stress and/or during rehydration stages. Results above will deepen our understanding of the molecular mechanisms of plant response to abiotic stress from the perspective of AS-network.
Related Materials