EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[Systemic mastocytosis].
PMID 25076308
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000639
Cladribine, European Pharmacopoeia (EP) Reference Standard
C10H12ClN5O3