EMAIL THIS PAGE TO A FRIEND
British Journal of Cancer
Dasatinib worsens the effect of cetuximab in combination with fractionated radiotherapy in FaDu- and A431-derived xenografted tumours.
PMID 25077442