EMAIL THIS PAGE TO A FRIEND
The Journal of General Virology
Epitope mapping of the 2009 pandemic and the A/Brisbane/59/2007 seasonal (H1N1) influenza virus haemagglutinins using mAbs and escape mutants.
PMID 25078301