EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Development and characterization of a novel, antimicrobial, sterile hydrogel dressing for burn wounds: single-step production with gamma irradiation creates silver nanoparticles and radical polymerization.
PMID 25079080
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282731
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%
C7H13NO4S