EMAIL THIS PAGE TO A FRIEND
Clinical Endocrinology
A longitudinal study of thyroid autoantibodies in pregnancy: the importance of test timing.
PMID 25079145
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57660
Iodine, purum p.a., ≥99.5% (RT)
I2