PloS one
Itm2a expression in the developing mouse first lower molar, and the subcellular localization of Itm2a in mouse dental epithelial cells.
PMID 25079563
Abstract
Itm2a is a type II transmembrane protein with a BRICHOS domain. We investigated the temporospatial mRNA and protein expression patterns of Itm2a in the developing lower first molar, and examined the subcellular localization of Itm2a in murine dental epithelial (mDE6) cells. From the initiation to the bud stage, the in situ and protein signals of Itm2a were not detected in either the dental epithelial or mesenchymal cells surrounding the tooth bud. However, at the bell stage, these signals of Itm2a were primarily observed in the inner enamel epithelium of the enamel organ. After the initiation of the matrix formation, strong signals were detected in ameloblasts and odontoblasts. Itm2a showed a punctate pattern in the cytoplasm of the mDE6 cells. The perinuclear-localized Itm2a displayed a frequent overlap with the Golgi apparatus marker, GM130. A tiny amount of Itm2a was colocalized with lysosomes and endoplasmic reticulum. Minimal or no overlap between the Itm2a-EGFP signals with the other organelle markers for endoplasmic reticulum, lysosome and mitochondria used in this study noted in the cytoplasm. These findings suggest that Itm2a may play a role in cell differentiation during odontogenesis, rather than during the initiation of tooth germ formation, and may be related to the targeting of proteins associated with enamel and dentin matrices in the secretory pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7295
Anti-GM130 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA021799
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA021178
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA021230
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
PRS3627
Anti-LAMP-2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
L0668
Anti-LAMP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA029100
Anti-LAMP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701987
Anti-LAMP2 antibody produced in rabbit
SAB2702058
Anti-LAMP2 antibody produced in rabbit
M6319
Anti-Mitofusin-2 (N-Terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1304456
ANTI-PDIA2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501845
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501851
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA053492
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA051692
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405897
Anti-PDIA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003230
Anti-PDIA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002645
Anti-PDIA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410582
Anti-PDIA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
D9026
Digoxigenin, analytical standard
C23H34O5
Y0000640
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
C23H34O5
WH0002801M1
Monoclonal Anti-GOLGA2 antibody produced in mouse, clone 2C6, purified immunoglobulin, buffered aqueous solution
SAB1402250
Monoclonal Anti-LAMP2 antibody produced in mouse, clone 2G10, purified immunoglobulin, buffered aqueous solution
SAB1405886
Anti-GOLGA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2107799
Anti-PDIA3 antibody produced in rabbit, affinity isolated antibody