EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Cytotoxic diterpenoids from Salvia yunnanensis.
PMID 25081107
Abstract
Forty-six abietane type diterpenoids possessing nine different fused ring systems were characterized from the roots of Salvia yunnanensis, six of which (salyunnanins A-F, 1-6) had different nor-abietane, homo-abietane, seco-abietane, and normal abietane architectures. Their structures were elucidated by comprehensive NMR and MS spectroscopic analyses. The inhibitory activities of these isolates against six human tumor lines were tested in vitro. Several of the compounds exhibited substantial cytotoxicity with IC50 values of 0.86-10.1μM.