EMAIL THIS PAGE TO A FRIEND
Transplant International : Official Journal of the European Society For Organ Transplantation
Uremia-associated immunological aging is stably imprinted in the T-cell system and not reversed by kidney transplantation.
PMID 25082296