EMAIL THIS PAGE TO A FRIEND
Journal of Neuroscience Research
Postconditioning mitigates cell death following oxygen and glucose deprivation in PC12 cells and forebrain reperfusion injury in rats.
PMID 25082329