EMAIL THIS PAGE TO A FRIEND
Cancer letters
Anti-cancer effect of tectochrysin in NSCLC cells through overexpression of death receptor and inactivation of STAT3.
PMID 25083589
Abstract
Phenolic compounds (flavonoids and phenolic acid derivatives) are the most important pharmacologically active ingredients, and these compounds could inhibit proliferation of human cancer cells by inducing of apoptotic cell death. Here we focused on the anticancer effects of tectochrysin on human non-small-cell lung cancer (NSCLC) cells and its mechanism of action. We analysed the activity of tectochrysin on NSCLC cells (A549 and NCI-H460) by use of Western blot analysis for major apoptotic proteins and death receptor expression. We also used EMSA for effects on STAT3 DNA binding activity. Tectochrysin (0-80 μM) suppressed the growth of A549 and NCI-H460 lung cancer cells by inducing of apoptotic cell death in a concentration dependent manner. Expression of DR3 and Fas as well as DR downstream pro-apoptotic proteins including cleaved caspase-3, cleaved caspase-8, cleaved caspase-9 and Bax were concomitantly increased, but the expression of anti-apoptotic proteins; Bcl-2 was decreased in both cancer cells. In addition, tectochrysin treatment also inhibited phosphorylation of STAT3 in A549 and NCI-H460 cells. However, deletion of DR3 and Fas by small interfering RNA significantly reversed tectochrysin-induced cell growth inhibitory effect as well as down regulation of STAT3 in A549 and NCI-H460 lung cancer cells. Pull down assay and docking model showed interaction of tectochrysin with STAT3. We propose that tectochrysin leads to apoptotic cell death in NSCLC cells through activation of DR3 and Fas expression via inhibition of STAT3 phosphorylation.