EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Ventromedial hypothalamus-specific Ptpn1 deletion exacerbates diet-induced obesity in female mice.
PMID 25083988
Abstract
Protein-tyrosine phosphatase 1B (PTP1B) regulates food intake (FI) and energy expenditure (EE) by inhibiting leptin signaling in the hypothalamus. In peripheral tissues, PTP1B regulates insulin signaling, but its effects on CNS insulin action are largely unknown. Mice harboring a whole-brain deletion of the gene encoding PTP1B (Ptpn1) are lean, leptin-hypersensitive, and resistant to high fat diet-induced (HFD-induced) obesity. Arcuate proopiomelanocortin (POMC) neuron-specific deletion of Ptpn1 causes a similar, but much milder, phenotype, suggesting that PTP1B also acts in other neurons to regulate metabolism. Steroidogenic factor-1-expressing (SF-1-expressing) neurons in the ventromedial hypothalamus (VMH) play an important role in regulating body weight, FI, and EE. Surprisingly, Ptpn1 deletion in SF-1 neurons caused an age-dependent increase in adiposity in HFD-fed female mice. Although leptin sensitivity was increased and FI was reduced in these mice, they had impaired sympathetic output and decreased EE. Immunohistochemical analysis showed enhanced leptin and insulin signaling in VMH neurons from mice lacking PTP1B in SF-1 neurons. Thus, in the VMH, leptin negatively regulates FI, promoting weight loss, whereas insulin suppresses EE, leading to weight gain. Our results establish a novel role for PTP1B in regulating insulin action in the VMH and suggest that increased insulin responsiveness in SF-1 neurons can overcome leptin hypersensitivity and enhance adiposity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1570428
2-Propanol, United States Pharmacopeia (USP) Reference Standard
C3H8O
PHR1072
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O
91237
2-Propanol, analytical standard
C3H8O
B5283 Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
R2655
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
49152
D-(+)-Glucose, tested according to Ph Eur
C6H12O6
PHR1000
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
1181302
Dextrose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
R1008 Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
A2556
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
R3155 Monoclonal Anti-Rabbit IgG, Native−Peroxidase antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
T1500
Testosterone, ≥98%
C19H28O2
T0100000
Testosterone, European Pharmacopoeia (EP) Reference Standard
C19H28O2
46923
Testosterone, VETRANAL, analytical standard
C19H28O2
Y0000342
Testosterone for impurity D identification, European Pharmacopoeia (EP) Reference Standard
C19H28O2
Y0000343
Testosterone for system suitability, European Pharmacopoeia (EP) Reference Standard
C19H28O2
T-037
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
C19H28O2