EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological and toxicological methods
Determination of receptor protein binding site specificity and relative binding strength using a time-resolved competition assay.
PMID 25084055
Abstract
Competitive binding assays can be used to decipher not only the binding kinetics of studied ligands but also the binding site preference. Such assays are an essential step in the characterization of radioligands. However, the currently used competition assays require high concentrations of usually expensive ligands and still provide only binding site preference. By employing the time-resolved competition assay presented in this paper, binding characteristics including binding site preference can be obtained using less ligand. To demonstrate the appropriateness of the time-resolved competition assay, we developed an assay in which the ligand binding was interrupted with a competitor. Experiments were performed on human carcinoma cell lines expressing epidermal growth factor receptor (EGFR). The targeting of the receptor was performed with radio-iodinated epidermal growth factor (EGF). The employed competitors involved either natural ligand transforming growth factor alpha (TGF-α) or anti-EGFR antibodies cetuximab and panitumumab targeting the same EGFR domain. Radio-iodinated EGF bound to EGFR was displaced with either low concentrations of cetuximab or high concentrations of panitumumab. In the case of TGF-α, we observed no competitive displacement of bound EGF at either high or low concentrations. When comparing the time-resolved competition assay with a manual competition assay, the resulting data of measured inhibition constants were in agreement. The results summarised in this study confirm the appropriateness of the time-resolved competition assay for assessing ligand binding properties. The assay has the potential to complement or replace conventional competition assays for determining binding site preference in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018530
Anti-EGFR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
WH0001956M2
Monoclonal Anti-EGFR antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
AMAB90816
Monoclonal Anti-EGFR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0815, purified immunoglobulin, buffered aqueous glycerol solution
E2760
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
E3138
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
E0532 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
PLA0101
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.