EMAIL THIS PAGE TO A FRIEND
The International journal of clinical and experimental hypnosis
Carriers of the COMT Met/Met allele have higher degrees of hypnotizability, provided that they have good attentional control: a case of gene-trait interaction.
PMID 25084618